missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.6%, ACS reagent, stabilized with amylene

Product Code. 10750862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10750862 500 mL EcoSafPak™ Glass Bottle 500mL
10424121 1 L Glass Bottle 1 L
10207283 2.5 L Glass Bottle 2.5 L
10532904 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10750862 Supplier Thermo Scientific Acros Supplier No. 406925000
  • ONLINE EXCLUSIVE PRICE
    33.40 €
    32.40 € / 500mL
    Save 1.00 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Decomposition Information >120°C
Density 1.3250g/mL
Quantity 500 mL
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range 99.6%
Specific Gravity 1.325
Titratable Acid 0.0003 meq/g max.
Linear Formula CH2Cl2
Vapor Pressure Vapor Pressure: 475hPa at 20°C
Viscosity 0.43 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Boiling Point 39.0°C to 40.0°C
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Dichloromethane, Stabilized with Amylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.