missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™

Product Code. 10354263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10354263 500 mL AcroSeal™ Glass Bottle 500mL
10487532 100 mL AcroSeal™ Glass Bottle 100mL
10387841 1 L AcroSeal™ Glass Bottle 1 L
10295661 2.5 L AcroSeal™ Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10354263 Supplier Thermo Scientific Acros Supplier No. 348465000
  • ONLINE EXCLUSIVE PRICE
    59.35 €
    57.60 € / 500mL
    Save 1.75 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Decomposition Information >120°C
Melting Point -97.0°C
Color Colorless
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Infrared Spectrum Authentic
Assay Percent Range 99.8% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH2Cl2
Quantity 500 mL
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Specific Gravity 1.325
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Stabilizer 50ppm (approx.) amylene
Viscosity 0.43 mPa.s (20°C)
Formula Weight 84.93
Vapor Pressure 475hPa at 20°C
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Physical Form Liquid
Chemical Name or Material Dichloromethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.