missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene

Product Code. 10205671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
Unit Size:
10 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10205671 2.5 L Glass Bottle 2.5 L
10793904 10 L Metal Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205671 Supplier Thermo Scientific Acros Supplier No. 354800025
  • ONLINE EXCLUSIVE PRICE
    157.00 €
    152.00 € / 2.5 L
    Save 5.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For peptide synthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Density 1.3250g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9%
Specific Gravity 1.325
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Water 0.005% max.
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Boiling Point 39.0°C to 40.0°C
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.9%
Chemical Name or Material Dichloromethane, Stabilized with Amylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.