missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, for residue and pestic. anal., stab. with amylene

Product Code. 10517631 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
missing translation for 'unitSize'
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10517631 1 L 1 L
10369270 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10517631 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 326600010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Decomposition Information >120°C
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 2ppm max.
Specific Gravity 1.325
Linear Formula CH2Cl2
Vapor Pressure Vapor Pressure: 475hPa at 20°C
Viscosity 0.43 mPa.s (20°C)
Beilstein 01, 60
Water 0.02% max. (K.F.)
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Chemical Name or Material Dichloromethane, ECD Tested, For Pesticide analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.