missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac diethylamine, MedChemExpress

Product Code. 16456298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456298 10 g 10g
16448887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 16456298 Proveedor Medchem Express N.º de proveedor HY15036A10g
 Restricted Item
Estimated Shipment: 30-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.

Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.

Especificaciones

Chemical Name or Material Diclofenac diethylamine
CAS 78213-16-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H22Cl2N2O2
Quantity 10 g
Solubility Information DMSO : 150 mg/mL (406.18 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl.CCNCC
Molecular Weight (g/mol) 369.29
Formula Weight 369.29
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.