missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicloxacillin Sodium hydrate, MedChemExpress
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Dicloxacillin Sodium hydrate (Dicloxacillin sodium salt monohydrate) is a narrow-spectrum β-Lactam antibiotic of the penicillin class, is used to treat infections caused by susceptible Gram-positive bacteria, active against beta-lactamase-producing organisms such as Staphylococcus aureus.
Specifications
Specifications
| Chemical Name or Material | Dicloxacillin Sodium hydrate |
| Physical Form | Solid |
| CAS | 13412-64-1 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18Cl2N3NaO6S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Dicloxacillin sodium salt monohydrate |
| Solubility Information | H2O : 25 mg/mL (48.99 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction