missing translation for 'onlineSavingsMsg'
Learn More

Dicumarol

Product Code. 15435819
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435819 5 g 5g
15445819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435819 Supplier Thermo Scientific Alfa Aesar Supplier No. J63634.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vitamin K reductase inhibitor possessing anticoagulant properties | CAS: 66-76-2 | C19H12O6 | 336.30 g/mol

Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

Solubility
Soluble in aqueous alkaline solutions, organic bases, 0.1 N NaOH (15 mg/ml), Pyridine (50 mg/ml), chloroform (slightly soluble), and benzene (slightly soluble). Insoluble in water, and alcohols.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.30
MDL Number MFCD00006857
InChI Key DOBMPNYZJYQDGZ-UHFFFAOYSA-N
Synonym dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
PubChem CID 54676038
ChEBI CHEBI:4513
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
SMILES OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12

Specifications

Melting Point 286°C to 289°C
Color White
Quantity 5 g
UN Number UN2811
Merck Index 14,3090
Solubility Information Soluble in aqueous alkaline solutions,organic bases,0.1 N NaOH (15mg/ml),Pyridine (50mg/ml),chloroform (slightly soluble),and benzene (slightly soluble). Insoluble in water,and alcohols.
Formula Weight 336.3
Physical Form Powder
Chemical Name or Material Dicumarol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicumarol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Dicoumarol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.