missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl peroxide, 98%

Product Code. 15406918
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406918 500 g 500g
15496908 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406918 Supplier Thermo Scientific Alfa Aesar Supplier No. H60442.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-43-3 | C18H22O2 | 270.372 g/mol

Dicumyl peroxide (DCP) is used:

  • in vulcanization of rubber.
  • as a cross linking agent in the synthesis of polylactic acid composite fibers.
  • in the preparation of polyethylene composites.
  • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicumyl peroxide (DCP) is used: • in vulcanization of rubber. • as a cross linking agent in the synthesis of polylactic acid composite fibers. • in the preparation of polyethylene composites. • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.

Solubility
Soluble in chloroform and most organic solvents. Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.372
MDL Number MFCD00036227
InChI Key XMNIXWIUMCBBBL-UHFFFAOYSA-N
Synonym dicumyl peroxide, cumene peroxide, cumyl peroxide, percumyl d, perkadox b, diisopropylbenzene peroxide, perkadox bc, perkadox sb, dicumenyl peroxide, peroxide, bis 1-methyl-1-phenylethyl
PubChem CID 6641
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)OOC(C)(C)C2=CC=CC=C2

Specifications

Melting Point 39°C to 41°C
Density 1.56
Linear Formula [C6H5C(CH3)2]2O2
Quantity 500 g
UN Number UN3110
Beilstein 2056090
Solubility Information Soluble in chloroform and most organic solvents. Insoluble in water.
Formula Weight 270.37
Percent Purity 98%
Chemical Name or Material Dicumyl peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.