missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl peroxide, 99%

Product Code. 10123051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123051 100 g Plastic bottle 100g
10536821 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123051 Supplier Thermo Scientific Acros Supplier No. 349881000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-43-3 | C18H22O2 | 270.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00036227
InChI Key XMNIXWIUMCBBBL-UHFFFAOYSA-N
Synonym dicumyl peroxide, cumene peroxide, cumyl peroxide, percumyl d, perkadox b, diisopropylbenzene peroxide, perkadox bc, perkadox sb, dicumenyl peroxide, peroxide, bis 1-methyl-1-phenylethyl
PubChem CID 6641
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)OOC(C)(C)C2=CC=CC=C2

Specifications

Melting Point 38.0°C to 41.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula [C6H5C(CH3)2]2O2
Quantity 100 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 270.37
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Dicumyl peroxide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicumyl peroxide
Signal Word
  • Danger||Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Organic peroxide Type E & F
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H242-Heating may cause a fire.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.