missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl phthalate, 99%

Product Code. 10433433 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
5g
This item is not returnable. View return policy

Product Code. 10433433

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84-61-7 | C20H26O4 | 330.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-61-7
Molecular Formula C20H26O4
Molecular Weight (g/mol) 330.42
MDL Number MFCD00003849
InChI Key VOWAEIGWURALJQ-UHFFFAOYSA-N
Synonym dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190
PubChem CID 6777
ChEBI CHEBI:34693
IUPAC Name dicyclohexyl benzene-1,2-dicarboxylate
SMILES C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3

Specifications

Melting Point 63.0°C
Color White
Density 207g/mL
Boiling Point 200.0°C to 235.0°C (4.0 mmHg)
Flash Point 207°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 330.42
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexyl phthalate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dicyclohexyl phthalate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.