missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Product Code. 10153051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153051 5 g Glass bottle 5g
10767772 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153051 Supplier Thermo Scientific Acros Supplier No. 348510050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-21-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00077209
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
PubChem CID 6673
SMILES C1C2C3CC4C(C3C1C5C2O5)O4

Specifications

Melting Point 185.0°C to 189.0°C
Color Beige-Yellow to White
Boiling Point 326.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 1.4g/L (20°C)
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less
compliance-icons
Product Identifier
  • Dicyclopentadiene diepoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.