missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylphenylsilane, 97+%

Product Code. 11400098
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400098 10 g 10g
11410098 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400098 Supplier Thermo Scientific Alfa Aesar Supplier No. B23684.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 775-56-4 | C11H18O2Si | 210.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-56-4
Molecular Formula C11H18O2Si
Molecular Weight (g/mol) 210.35
MDL Number MFCD00048541
InChI Key MNFGEHQPOWJJBH-UHFFFAOYSA-N
Synonym methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane
PubChem CID 69891
IUPAC Name diethoxy-methyl-phenylsilane
SMILES CCO[Si](C)(OCC)C1=CC=CC=C1

Specifications

Boiling Point 117°C to 118°C (30mmHg)
Quantity 10 g
Beilstein 2938396
Flash Point 88°C (190°F)
Formula Weight 210.35
Percent Purity ≥97%
Refractive Index 1.4696
Sensitivity Moisture sensitive
Density 0.963
Chemical Name or Material Diethoxymethylphenylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.