missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3

Product Code. 10377901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377901 5 g Glass bottle 5g
10387181 25 g Glass bottle 25g
10492835 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377901 Supplier Thermo Scientific Acros Supplier No. 346930050
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15110-17-5 | C11H25O5P | 268.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15110-17-5
Molecular Formula C11H25O5P
Molecular Weight (g/mol) 268.29
MDL Number MFCD00800259
InChI Key WKPXMGJOGYEGNW-UHFFFAOYSA-N
Synonym diethyl 3,3-diethoxypropyl phosphonate, diethyl 3,3-diethoxypropylphosphonate, phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester, 3-diethoxyphosphoryl-1,1-diethoxy-propane, 3-diethoxyphosphoryl propanaldehyde diethyl acetal, phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester
PubChem CID 328856
IUPAC Name 3-diethoxyphosphoryl-1,1-diethoxypropane
SMILES CCOC(CCP(=O)(OCC)OCC)OCC

Specifications

Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4320
Quantity 5 g
Stabilizer 1% max. Na2CO3
Formula Weight 268.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl (3, 3-diethoxypropyl)phosphonate, Stabilized with max.1% (w/w) Na2CO3
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl (3,3-diethoxypropyl)phosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.