missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3

Product Code. 10387181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387181 25 g Glass bottle 25g
10377901 5 g Glass bottle 5g
10492835 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387181 Supplier Thermo Scientific Acros Supplier No. 346930250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 15110-17-5 | C11H25O5P | 268.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 15110-17-5
Molecular Formula C11H25O5P
Molecular Weight (g/mol) 268.29
MDL Number MFCD00800259
InChI Key WKPXMGJOGYEGNW-UHFFFAOYSA-N
Synonym diethyl 3,3-diethoxypropyl phosphonate, diethyl 3,3-diethoxypropylphosphonate, phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester, 3-diethoxyphosphoryl-1,1-diethoxy-propane, 3-diethoxyphosphoryl propanaldehyde diethyl acetal, phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester
PubChem CID 328856
IUPAC Name 3-diethoxyphosphoryl-1,1-diethoxypropane
SMILES CCOC(CCP(=O)(OCC)OCC)OCC

Specifikationer

Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4320
Quantity 25 g
Stabilizer 1% max. Na2CO3
Formula Weight 268.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl (3, 3-diethoxypropyl)phosphonate, Stabilized with max.1% (w/w) Na2CO3
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Diethyl (3,3-diethoxypropyl)phosphonate
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.