missing translation for 'onlineSavingsMsg'
Learn More

Diethyl aminomethylenemalonate, 98%

Product Code. 11388286
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11388286

Brand: Thermo Scientific Alfa Aesar L15511.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6296-99-7 | C8H13NO4 | 187.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6296-99-7
Molecular Formula C8H13NO4
Molecular Weight (g/mol) 187.195
MDL Number MFCD01075695
InChI Key WDTWMEZMHUEZKH-UHFFFAOYSA-N
Synonym diethyl 2-aminomethylene malonate, diethyl aminomethylenemalonate, 1,3-diethyl 2-aminomethylidene propanedioate, propanedioic acid, aminomethylene-, diethyl ester, diethyl aminomethylene malonate, amino-methylene malonic acid, diethyl ester, acmc-1b66v, ksc494c4d, diethyl amino-methylenemalonate
PubChem CID 226751
IUPAC Name diethyl 2-(aminomethylidene)propanedioate
SMILES CCOC(=O)C(=CN)C(=O)OCC

Specifications

Melting Point 60°C to 65°C
Boiling Point 114°C (1 mmHg)
Quantity 1 g
Beilstein 1708651
Formula Weight 187.2
Percent Purity 98%
Chemical Name or Material Diethyl aminomethylenemalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.