missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl benzamidomalonate, 98+%

Product Code. 11387675
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387675 5 g 5g
11397675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387675 Supplier Alfa Aesar™ Supplier No. L02371.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16798-45-1
Molecular Formula C14H17NO5
Molecular Weight (g/mol) 279.29
MDL Number MFCD00015160
InChI Key UQFFIFUYIQXSPD-UHFFFAOYSA-N
Synonym diethyl benzamidomalonate, diethyl 2-benzamidomalonate, 1,3-diethyl 2-phenylformamido propanedioate, diethyl-n-benzoylamino malonate, benzamidomalonic acid, diethyl ester, diethyl benzoylaminomalonate, diethyl 2-phenylcarbonylamino propane-1,3-dioate, diethyl benzoylamino malonate, ethyl benzaminomalonate, diethylbenzamidomalonate
PubChem CID 66790
IUPAC Name diethyl 2-benzamidopropanedioate
SMILES CCOC(=O)C(C(=O)NC1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 61°C to 65°C
Quantity 5 g
Beilstein 2817007
Formula Weight 279.29
Percent Purity ≥98%
Chemical Name or Material Diethyl benzamidomalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.