missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isocyanomethylphosphonate, 97%

Product Code. 10620893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10620893 1 mL Glass bottle 1mL
10647094 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10620893 Supplier Thermo Scientific Acros Supplier No. 369240010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41003-94-5 | C6H12NO3P | 177.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41003-94-5
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00042951
InChI Key ICURVXBGGDVHDT-UHFFFAOYSA-N
Synonym diethyl isocyanomethylphosphonate, diethylisocyanomethylphosphonate, acmc-20al8q, diethoxycarbonyl methanisocyanide, diethyl isocyanomethyl phosphonate, diethoxyphosphinylmethyl isocyanide, isocyanomethylphosphonic acid diethyl ester, isocyanomethylphosphononic acid diethyl ester, 1-ethoxy isocyanomethyl phosphoryl oxyethane
PubChem CID 2734699
IUPAC Name 1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane
SMILES CCOP(=O)(C[N+]#[C-])OCC

Specifications

Color Colorless to Yellow
Density 1.1050g/mL
Boiling Point 84°C to 87°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CNCH2P(O)(OC2H5)2
Refractive Index 1.4310 to 1.4340 (20°C, 589nm)
Quantity 1 mL
Fieser 14,135
Specific Gravity 1.105
Formula Weight 177.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl isocyanomethylphosphonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl isocyanomethylphosphonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.