missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phthalate, 99%

Product Code. 10714801 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10714801 2.5 L Glass bottle 2.5 L
10376080 25 mL Glass bottle 25mL
10569410 1 L Glass bottle 1 L
10443825 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10714801 Supplier Thermo Scientific Acros Supplier No. 114520025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Density 1.1180g/mL
Boiling Point 298.0°C to 299.0°C
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5010 to 1.5030
Quantity 2.5 L
Beilstein 09,798
Merck Index 15,7483
Specific Gravity 1.118
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride
Viscosity 12 mPa.s (20°C)
Formula Weight 222.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl phthalate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.