missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate, TRC

Product Code. 30012499
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012499 100 g 100g
30011852 500 g 500g
17918324 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012499 Supplier Toronto Research Chemicals Supplier No. TRCD444770100G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12 H14 O4
Molecular Weight (g/mol) 222.24
Synonym 1,2-Benzenedicarboxylic acid, 1,2-diethyl ester, 1,2-Benzenedicarboxylic acid, diethyl ester (9CI), Phthalic acid, diethyl ester (6CI,8CI), 1,2-Benzodioic acid diethyl ester, 1,2-Diethyl benzene-1,2-dicarboxylate, Anozol, DEP, Diethyl 1,2-benzenedicarboxylate, Diethyl phthalate, Ethyl phthalate, NSC 8905, Neantine, Palatinol A, Phthalol, Placidol E, Solvanol, Unimoll DA, o-Benzenedicarboxylic acid diethyl ester, o-Bis(ethoxycarbonyl)benzene, Phthalic acid, bis-ethyl ester
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)c1ccccc1C(=O)OCC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3
Formula Weight 222.0892
Percent Purity >95%
Chemical Name or Material Diethyl phthalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.