missing translation for 'onlineSavingsMsg'
Learn More

Diethyl terephthalate, 95%

Product Code. p-7025743
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11439357

Brand: Thermo Scientific Alfa Aesar B21401.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-09-9 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 44°C to 46°C
Density 1.1
Quantity 10 g
Formula Weight 222.24
Percent Purity 95%
Chemical Name or Material Diethyl terephthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.