missing translation for 'onlineSavingsMsg'
Learn More

Diethylamino hydroxybenzoyl hexyl benzoate, MedChemExpress

Product Code. 16491586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491586 500 mg 500mg
16465236 100 mg 100mg
16411139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491586 Supplier Medchem Express Supplier No. HY109656

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber.

Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber.

Specifications

Chemical Name or Material Diethylamino hydroxybenzoyl hexyl benzoate
CAS 302776-68-7
Color White
Purity Grade Notes Research
Molecular Formula C24H31NO4
Quantity 500 mg
Synonym DHHB
Solubility Information DMSO : ≥ 50 mg/mL (125.78 mM)
SMILES O=C(OCCCCCC)C1=CC=CC=C1C(C2=CC=C(N(CC)CC)C=C2O)=O
Molecular Weight (g/mol) 397.51
Formula Weight 397.51
Percent Purity 99.88%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.