missing translation for 'onlineSavingsMsg'
Learn More

Diethylstilbestrol, 99%

Product Code. 10358340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10358340 10 g 10g
10297113 1 g 1g
10124690 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10358340 Supplier Thermo Scientific Acros Supplier No. 204130100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-53-1 | C18H20O2 | 268.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-53-1
Molecular Formula C18H20O2
Molecular Weight (g/mol) 268.36
MDL Number MFCD00002373
InChI Key RGLYKWWBQGJZGM-ISLYRVAYSA-N
Synonym diethylstilbestrol, stilbestrol, stilboestrol, distilbene, agostilben, estrobene, estromenin, stilboestroform, antigestil, palestrol
PubChem CID 448537
ChEBI CHEBI:41922
SMILES CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 169°C to 175°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 06, III, 5619
Merck Index 15, 3148
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in ethylether,ethanol,chloroform,aceton,e,methanol,arachis oil,olive oil
IUPAC Name 4-[(E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol
Formula Weight 268.34
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diethylstilbestrol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diethylstilbestrol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360D-May damage the unborn child.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.