missing translation for 'onlineSavingsMsg'
Learn More

Diethyltin dichloride

Product Code. p-7080325
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394839 0.5 g 0.5g
11304849 2 g 2g
11314849 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394839 Supplier Thermo Scientific Alfa Aesar Supplier No. 071155.02
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 866-55-7 | C4H10Cl2Sn | 247.734 g/mol

Used as organic intermediates and active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-55-7
Molecular Formula C4H10Cl2Sn
Molecular Weight (g/mol) 247.734
MDL Number MFCD00013609
InChI Key YFAXVVMIXZAKSR-UHFFFAOYSA-L
Synonym diethyltin dichloride, diethyldichlorotin, diethyltindichloride, dichlorodiethyltin, diethyldichlorostannane, tin, dichlorodiethyl, stannane, dichlorodiethyl, diethyltin chloride, tin, diethyl-, dichloride, unii-pga900d045
PubChem CID 91491
IUPAC Name dichloro(diethyl)stannane
SMILES CC[Sn](CC)(Cl)Cl

Specifications

Melting Point 84°C
Color White
Boiling Point 220°C
Linear Formula (C2H5)2SnCl2
Quantity 0.5 g
Sensitivity Moisture sensitive
Solubility Information Partly soluble in water((0.023g/L) (25°C),Calc).
Formula Weight 247.72
Physical Form Crystalline
Chemical Name or Material Diethyltin dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.