missing translation for 'onlineSavingsMsg'
Learn More

Diethyltoluamide, MedChemExpress

Product Code. 16422299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422299 Supplier Medchem Express Supplier No. HYB0978
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diethyltoluamide is the most common active ingredient in insect repellents. It is intended to provide protection against mosquitoes, ticks, fleas, chiggers, leeches, and many other biting insects.

Diethyltoluamide is the most common active ingredient in insect repellents. It is intended to provide protection against mosquitoes, ticks, fleas, chiggers, leeches, and many other biting insects.

Specifications

Chemical Name or Material Diethyltoluamide
Physical Form Liquid
CAS 134-62-3
Color Light Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C12H17NO
For Use With (Application) COVID-19-immunoregulation
Synonym DEET N,N-Diethyl-m-toluamide
Solubility Information DMSO : ≥ 100 mg/mL (522.82 mM) ∣H2O : 2 mg/mL (10.46 mM; Need ultrasonic)
SMILES O=C(N(CC)CC)C1=CC=CC(C)=C1
Molecular Weight (g/mol) 191.27
Formula Weight 191.27
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.