missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diflufenican metabolite AE B107137 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Diflufenican Metabolite AE B107137 |
| CAS | 36701-89-0 |
| Analyte or Component Names | Diflufenican Metabolite AE B107137 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H8 F3 N O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Diflufenican Metabolite AE B107137, 2-[3-(Trifluoromethyl)phenoxy]-3-pyridinecarboxylic acid, 2-(3-Trifluoromethylphenoxy)-3-pyridinecarboxylic acid, 2-(3-Trifluoromethylphenoxy)nicotinic acid, 3-Pyridinecarboxylic acid, 2-[3-(trifluoromethyl)phenoxy]- |
| InChI Formula | InChI=1S/C13H8F3NO3/c14-13(15,16)8-3-1-4-9(7-8)20-11-10(12(18)19)5-2-6-17-11/h1-7H,(H,18,19) |
| SMILES | OC(=O)c1cccnc1Oc2cccc(c2)C(F)(F)F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?