missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic anhydride, 97%

Product Code. 10060820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10060820 25 g Glass bottle 25g
10685912 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10060820 Supplier Thermo Scientific Acros Supplier No. 114670250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4480-83-5 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4480-83-5
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00006677
InChI Key PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Synonym diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
PubChem CID 78232
IUPAC Name 1,4-dioxane-2,6-dione
SMILES C1C(=O)OC(=O)CO1

Specifications

Melting Point 90°C to 95°C
Color Cream to White
Boiling Point 240°C to 241°C
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 25 g
Beilstein 19, 153
Solubility Information Solubility in water: soluble
Formula Weight 116.07
Percent Purity 0.97
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Diglycolic anhydride, 0.97
Show More Show Less
compliance-icons
Product Identifier
  • Diglycolic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.