missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7520-SP
This item is not returnable.
View return policy
Description
Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-Species samples. It is validated for Immunohistochemistry.
Specifications
| Digoxigenin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| DIG | |
| KLH-coupled Digoxigenin | |
| 25 μg | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 611621 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Digoxigenin labeled proteins, nucleic acids and Digoxigenin conjugated primary and secondary antibodies. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction