missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Digoxin Recombinant Rabbit Monoclonal Antibody (PSH04-40)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556507
This item is not returnable.
View return policy
Description
Digoxin Recombinant Monoclonal Antibody for Western Blot, IHC (F), IHC (P), IP, ELISA, DB
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
| Digoxin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| PSH04-40 | |
| Unconjugated | |
| C41H64O14; DGX; Lanoxin | |
| DIG-OVA. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction