missing translation for 'onlineSavingsMsg'
Learn More

Dihydroformononetin, TRC

Product Code. 30133159
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133159 50 mg 50mg
30137616 10 mg 10mg
30005772 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133159 Supplier Toronto Research Chemicals Supplier No. TRCD45333850MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4626-22-6
Molecular Formula C16 H14 O4
Molecular Weight (g/mol) 270.281
IUPAC Name 7-hydroxy-3-(4-methoxyphenyl)chroman-4-one
SMILES COc1ccc(cc1)C2COc3cc(O)ccc3C2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14O4/c1-19-12-5-2-10(3-6-12)14-9-20-15-8-11(17)4-7-13(15)16(14)18/h2-8,14,17H,9H2,1H3
Formula Weight 270.0892
Chemical Name or Material Dihydroformononetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.