missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1.1M in cyclohexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals

Product Code. 16605905
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16605905 100 mL 100mL
16695895 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16605905 Supplier Thermo Scientific Alfa Aesar Supplier No. H37108.AE
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
Molecular Formula C8H18Al
Molecular Weight (g/mol) 141.21
InChI Key SIPUZPBQZHNSDW-UHFFFAOYSA-N
IUPAC Name bis(2-methylpropyl)alumanyl
SMILES CC(C)C[Al]CC(C)C

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.