missing translation for 'onlineSavingsMsg'
Learn More

Dimethoate 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30323637
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323637 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323637 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010335ME
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethoate
CAS 60-51-5
Analyte or Component Names Dimethoate
Shipping Condition Room Temperature
Molecular Formula C5 H12 N O3 P S2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphorodithioic acid, O,O-dimethyl S-[2-(methylamino)-2-oxoethyl] ester, Phosphorodithioic acid, O,O-dimethyl S-(methylcarbamoylmethyl) ester (6CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 2-mercapto-N-methylacetamide (8CI), 8014 Bis HC, Aadimethoal, Agrithoate 40EC, American Cyanamid 12,880, BI 58, Bi 58 Nowy, Bi 58 Nowy 400EC, CL 12880, Cerox, Chloromezyl, Cygon, Cygon 2E, Cygon 400, Cygon 4E, Cygon Insecticide, Danadim, Daphene, De-fend, Dimate, Dimate (insecticide), Dimethoate, Dimeton, Dimevur, Dimezyl, Dimezyl 400EC, Ditimur 40, Dudu Ethoate, ENT 24650, Experimental Insecticide 12880, FIP, Fosfatox R, Fosfotox, Fosfotox R, Fosfotox R 35, Fostion MM, Laition, Lurgo, O,O-Dimethyl S-(N-methylcarbamoylmethyl) dithiophosphate, O,O-Dimethyl S-(N-methylcarbamoylmethyl) phosphorodithioate, PEI 75, Perfekthion, Perfekthion S, Phosphamid, Phosphamide, Racusan, Rogor, Rogor 20L, Rogor 40, Rogor L, Rogor P, Roxion, S-Methylcarbamoylmethyl O,O-dimethyl phosphorodithioate, Sinoratox, Sistemin, Super Ethoate, Systemin, Systemin (pesticide), Systoate, Tafgor 40EC, Tara, Tara 909, Tiomet
InChI Formula InChI=1S/C5H12NO3PS2/c1-6-5(7)4-12-10(11,8-2)9-3/h4H2,1-3H3,(H,6,7)
SMILES CNC(=O)CSP(=S)(OC)OC
IUPAC Name 2-dimethoxyphosphinothioylsulfanyl-N-methylacetamide
Solvent or Matrix Methanol
Molecular Weight (g/mol) 229.26
Quantity 1 mL
Formula Weight 228.9996 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.