missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3-nitrophthalate, 98%

Product Code. 11316976
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316976 25 g 25g
11306976 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316976 Supplier Thermo Scientific Alfa Aesar Supplier No. L06715.14
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13365-26-9 | C10H9NO6 | 239.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13365-26-9
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00017184
InChI Key MLQMIKSBTAZNBK-UHFFFAOYSA-N
PubChem CID 249912
IUPAC Name dimethyl 3-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 67°C to 70°C
Quantity 25 g
Beilstein 1885570
Synonym dimethyl 3-nitrophthalate, 3-nitrophthalic acid dimethyl ester, 3-nitro-phthalic acid dimethyl ester, 1,2-benzenedicarboxylic acid, 3-nitro-, 1,2-dimethyl ester, 1,2-dimethyl 3-nitrophthalate, dimethyl nitrophthalate, dimethyl3-nitrophthalate, maybridge1_000481, acmc-20al75, methyl 2-methoxycarbonyl-3-nitrobenzoate
Formula Weight 239.19
Percent Purity 98%
Chemical Name or Material Dimethyl 3-nitrophthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.