missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3-nitrophthalate, 98%

Product Code. p-7044488
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11306976

Brand: Thermo Scientific Alfa Aesar L06715.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13365-26-9 | C10H9NO6 | 239.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13365-26-9
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00017184
InChI Key MLQMIKSBTAZNBK-UHFFFAOYSA-N
PubChem CID 249912
IUPAC Name dimethyl 3-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 67°C to 70°C
Quantity 5 g
Beilstein 1885570
Synonym dimethyl 3-nitrophthalate, 3-nitrophthalic acid dimethyl ester, 3-nitro-phthalic acid dimethyl ester, 1,2-benzenedicarboxylic acid, 3-nitro-, 1,2-dimethyl ester, 1,2-dimethyl 3-nitrophthalate, dimethyl nitrophthalate, dimethyl3-nitrophthalate, maybridge1_000481, acmc-20al75, methyl 2-methoxycarbonyl-3-nitrobenzoate
Formula Weight 239.19
Percent Purity 98%
Chemical Name or Material Dimethyl 3-nitrophthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.