missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (4-fluorobenzyl)malonate, 97%

Product Code. 11461170
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461170 5 g 5g
11451170 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461170 Supplier Thermo Scientific Alfa Aesar Supplier No. H26310.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 252938-22-0 | C12H13FO4 | 240.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252938-22-0
Molecular Formula C12H13FO4
Molecular Weight (g/mol) 240.23
MDL Number MFCD01934558
InChI Key WDFUURBBGZTSIA-UHFFFAOYSA-N
Synonym dimethyl 4-fluorobenzyl malonate, dimethyl 2-4-fluorobenzyl malonate, 1,3-dimethyl 2-4-fluorophenyl methyl propanedioate, acmc-1cpd2, dimethyl 4-fluorobenzylmalonate, 2-4-fluorobenzyl-malonic acid dimethyl ester, dimethyl 2-4-fluorophenyl methyl propanedioate, dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate
PubChem CID 2781576
SMILES COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC

Specifications

Quantity 5 g
IUPAC Name dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Formula Weight 240.23
Percent Purity 97%
Chemical Name or Material Dimethyl (4-fluorobenzyl)malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.