missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-aminoisophthalate, 98%

Product Code. 11455636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455636 100 g 100g
11445636 25 g 25g
11465636 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455636 Supplier Thermo Scientific Alfa Aesar Supplier No. B20761.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-27-4 | C10H11NO4 | 209.201 g/mol

Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

Solubility
Soluble in Chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-27-4
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008435
InChI Key DEKPYXUDJRABNK-UHFFFAOYSA-N
PubChem CID 66831
IUPAC Name dimethyl 5-aminobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)N)C(=O)OC

Spezifikation

Melting Point 178°C to 182°C
Quantity 100 g
Beilstein 2116565
Synonym dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate
Solubility Information Soluble in Chloroform.
Formula Weight 209.2
Percent Purity 98%
Chemical Name or Material Dimethyl 5-aminoisophthalate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.