missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-aminoisophthalate, 98%

Product Code. p-7025333
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11445636

Brand: Thermo Scientific Alfa Aesar B20761.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-27-4 | C10H11NO4 | 209.201 g/mol

Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

Solubility
Soluble in Chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-27-4
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008435
InChI Key DEKPYXUDJRABNK-UHFFFAOYSA-N
PubChem CID 66831
IUPAC Name dimethyl 5-aminobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)N)C(=O)OC

Specifications

Melting Point 178°C to 182°C
Quantity 25 g
Beilstein 2116565
Synonym dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate
Solubility Information Soluble in Chloroform.
Formula Weight 209.2
Percent Purity 98%
Chemical Name or Material Dimethyl 5-aminoisophthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.