missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-bromoisophthalate, 98%

Product Code. 11410150
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410150 1 g 1g
11420150 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410150 Supplier Thermo Scientific Alfa Aesar Supplier No. H50629.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51760-21-5 | C10H9BrO4 | 273.082 g/mol

Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51760-21-5
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00078709
InChI Key QUJINGKSNJNXEB-UHFFFAOYSA-N
Synonym dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
PubChem CID 103954
IUPAC Name dimethyl 5-bromobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 273.08
Percent Purity 98%
Chemical Name or Material Dimethyl 5-bromoisophthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.