missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-hydroxyisophthalate, 98%

Product Code. 11407164
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407164 10 g 10g
11417164 50 g 50g
11427164 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407164 Supplier Thermo Scientific Alfa Aesar Supplier No. B21565.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13036-02-7 | C10H10O5 | 210.19 g/mol

Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13036-02-7
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.19
MDL Number MFCD00134367
InChI Key DOSDTCPDBPRFHQ-UHFFFAOYSA-N
PubChem CID 83065
IUPAC Name dimethyl 5-hydroxybenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(O)=C1)C(=O)OC

Specifications

Melting Point 162°C to 164°C
Quantity 10 g
Beilstein 2650120
Synonym dimethyl 5-hydroxyisophthalate, 5-hydroxyisophthalic acid dimethyl ester, 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester, dimethyl5-hydroxyisophthalate, timtec-bb sbb000491, 5-hydroxy-dimethylisophthalate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester, dimethyl-5-hydroxy-benzene-1,3-dicarbonate, methyl 5-hydroxy-3-methoxycarbonyl benzoate
Solubility Information Soluble in water.
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material Dimethyl 5-hydroxyisophthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.