missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl aminoterephthalate, 99%

Product Code. 11493214
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493214 250 g 250g
11483214 50 g 50g
11473214 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493214 Supplier Thermo Scientific Alfa Aesar Supplier No. A10567.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5372-81-6 | C10H11NO4 | 209.201 g/mol

Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5372-81-6
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008427
InChI Key DSSKDXUDARIMTR-UHFFFAOYSA-N
Synonym dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
PubChem CID 79336
IUPAC Name dimethyl 2-aminobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)N

Specifications

Melting Point 130°C to 134°C
Quantity 250 g
Beilstein 2808505
Solubility Information Insoluble in water.
Formula Weight 209.2
Percent Purity 99%
Chemical Name or Material Dimethyl aminoterephthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.