missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isophthalate, 98%

Product Code. 11490228
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490228 100 g 100g
11400238 500 g 500g
11410238 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490228 Supplier Thermo Scientific Alfa Aesar Supplier No. B23779.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Melting Point 67°C to 70°C
Density 1.194
Boiling Point 282°C
Flash Point 138°C (280°F)
Odor Mild
Quantity 100 g
Beilstein 1912251
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material Dimethyl isophthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.