missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl naphthalene-2,6-dicarboxylate, 99+%

Product Code. p-7047133
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11338636

Brand: Thermo Scientific Alfa Aesar L17689.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 840-65-3 | C14H12O4 | 244.246 g/mol

Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 840-65-3
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00004100
InChI Key GYUVMLBYMPKZAZ-UHFFFAOYSA-N
Synonym dimethyl 2,6-naphthalenedicarboxylate, 2,6-dicarbomethoxynaphthalene, 2,6-naphthalenedicarboxylic acid, dimethyl ester, unii-x4q5t8rs0g, dimethyl-2,6-naphthalenedicarboxylate, 2,6-naphthalenedicarboxylic acid dimethyl ester, x4q5t8rs0g, 2,6-naphthalene dicarboxylic acid, dimethyl ester, dimethylnaphthalene-2,6-dicarboxylate, 2,6-naphthalenedicarboxylic acid, 2,6-dimethyl ester
PubChem CID 61225
IUPAC Name dimethyl naphthalene-2,6-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)C(=O)OC

Specifications

Melting Point 187°C to 193°C
Flash Point 232°C (449°F)
Quantity 5 g
Beilstein 987259
Solubility Information Soluble in hot toluene (very faint turbidity). Insoluble in water.
Formula Weight 244.25
Percent Purity ≥99%
Chemical Name or Material Dimethyl naphthalene-2,6-dicarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.