missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminobenzaldehyde R6 EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12900404 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12900404 1 mL Ampoule 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12900404 Supplier Fisher Chemical Supplier No. RP 062 100
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 14214-32-5
Packaging Ampoule
Molecular Formula C16H18N2O3
Quantity 1 mL
Synonym difenoxuron, difenoxurone, lironion, pinoran, caswell no. 551c, difenoxuron bsi:iso, epa pesticide chemical code 551400, n-4-4-methoxyphenoxy phenyl-n,n-dimethylurea, 1,1-dimethyl-3-4-4-methoxyphenoxy phenyl urea, 3-4-4-methoxyphenoxy phenyl-1,1-dimethylurea
InChI Key AMVYOVYGIJXTQB-UHFFFAOYSA-N
SMILES CN(C)C(=O)NC1=CC=C(C=C1)OC2=CC=C(C=C2)OC
IUPAC Name 3-[4-(4-methoxyphenoxy)phenyl]-1,1-dimethylurea
Molecular Weight (g/mol) 286.331
PubChem CID 26576
compliance-icons
Product Identifier
  • Dimethylaminobenzaldehyde solution R6. European Pharmacopoeia Ref: 1029803 (RP 062)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.