missing translation for 'onlineSavingsMsg'
Learn More

Dinitrodurene, 97%, Thermo Scientific™

Product Code. 15259616 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15259616 5 g 5g
15249616 1 g 1g
15269616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15259616 Supplier Thermo Scientific Acros Supplier No. 117000050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5465-13-4
InChI Key AEPQXGFMAZTUEA-UHFFFAOYSA-N
Synonym 1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
PubChem CID 79593
IUPAC Name 1,2,4,5-tetramethyl-3,6-dinitrobenzene
SMILES CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C

Specifications

Melting Point 208.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C10H12N2O4
Linear Formula (CH3)4C6(NO2)2
Quantity 5 g
Beilstein 05,433
Molecular Weight (g/mol) 224.22
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material Dinitrodurene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dinitrodurene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.