missing translation for 'onlineSavingsMsg'
Learn More

Dinitrodurene, 97%, Thermo Scientific™

Product Code. p-7077146 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15249616

Brand: Thermo Scientific Acros 117000010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5465-13-4
InChI Key AEPQXGFMAZTUEA-UHFFFAOYSA-N
Synonym 1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
PubChem CID 79593
IUPAC Name 1,2,4,5-tetramethyl-3,6-dinitrobenzene
SMILES CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C

Specifications

Melting Point 208.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C10H12N2O4
Linear Formula (CH3)4C6(NO2)2
Quantity 1 g
Beilstein 05,433
Molecular Weight (g/mol) 224.22
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material Dinitrodurene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dinitrodurene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.