missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl phthalate, mixture of isomers, 96%

Product Code. 11465303
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11465303

Brand: Thermo Scientific Alfa Aesar A12871.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
IUPAC Name dinonyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Density 0.97
Flash Point 216°C (421°F)
Quantity 250 g
Beilstein 1916263
Formula Weight 418.62
Percent Purity 98%
Chemical Name or Material Dinonyl phthalate, mixture of isomers

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.