missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00 € - 463.00 €
Specifications
| Antigen | DIO3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633612
|
Novus Biologicals
NBP2-92482-0.02ml |
0.02 mL |
193.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671822
|
Novus Biologicals
NBP2-92482-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DIO3 Polyclonal antibody specifically detects DIO3 in Human samples. It is validated for Western BlotSpecifications
| DIO3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1735 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-170 of human DIO3 (NP_001353.4). HFLGRRRRGQPEPEVELNSEGEEVPPDDPPICVSDDNRLCTLASLKAVWHGQKLDFFKQAHEGGPAPNSEVVLPDGFQSQHILDYAQGNRPLVLNFGSCTU | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title