missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl Terepthalate, TRC

Product Code. 17978124
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978124 10 g 10g
17949184 1 g 1g
17968524 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17978124 Supplier Toronto Research Chemicals Supplier No. TRCD48185010G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6422-86-2
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym Terephthalic Acid Bis(2-ethylhexyl) Ester, 1,4-Benzenedicarboxylic acid, 1,4-bis(2-ethylhexyl) ester, 1,4-Benzenedicarboxylic acid, bis(2-ethylhexyl) ester (9CI), Terephthalic acid, bis(2-ethylhexyl) ester (6CI,7CI,8CI), 168SG, ADK Cizer D 810, Bis(2-ethylhexyl) terephthalate, DOTP, Di-(2-ethylhexyl) terephthalate, Dioctyl terephthalate, Eastman 168, Eastman 168SG, Eastman TM 168, HCCFlex SP 390, JP 110, Kodaflex DOTP, NEO-T, Palatinol DOTP, Plasticizer 168, UN 488
IUPAC Name bis(2-ethylhexyl) benzene-1,4-dicarboxylate
SMILES CCCCC(CC)COC(=O)c1ccc(cc1)C(=O)OCC(CC)CCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C24H38O4/c1-5-9-11-19(7-3)17-27-23(25)21-13-15-22(16-14-21)24(26)28-18-20(8-4)12-10-6-2/h13-16,19-20H,5-12,17-18H2,1-4H3
Formula Weight 390.277
Chemical Name or Material Terephthalic acid, bis(2-ethylhexyl) ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.