missing translation for 'onlineSavingsMsg'
Learn More

Diosmin, TRC

Product Code. 30138577
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138577 10 g 10g
30008800 1 g 1g
30009369 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138577 Supplier Toronto Research Chemicals Supplier No. TRCD48520010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-27-4
Molecular Formula C28 H32 O15
Molecular Weight (g/mol) 608.54
Synonym Diosmin, 7-[[6-O-(6-Deoxy-α-l-mannopyranosyl)-β-d-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, 3',5,7-Trihydroxy-4'-methoxyflavone 7-O-rutinoside, 3',5-Dihydroxyl-4'-methoxylflavanone-7-O-α-L-rhamnosyl (1→6)-β-D-glucopyranoside, 7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosyl(1→6)-glucoside, Diosmetin 7-O-rutinoside, Diosmetin 7-O-α-L-rhamnopyranosyl-(1→6)-β-D-glucopyranoside, Diosmetin 7-rutinoside, Diosmetin 7-β-rutinoside, Diosmil, Diosmine, Diosven, Dioven, Diovenor, Flebosmil, Flebosten, Hemerven, Insuven, Litosmil, Tovene, Varinon, Ven-Detrex, Venex 500, Venosmine
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C28H32O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-8,10,19,21-30,32-37H,9H2,1-2H3/t10-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1
Formula Weight 608.1741
Percent Purity >95%
Chemical Name or Material Diosmin (Barosmin)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.