missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl chlorophosphate, 98%

Product Code. 10112960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10112960 100 mL Glass bottle 100mL
10723661 25 mL Glass bottle 25mL
10206873 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10112960 Supplier Thermo Scientific Acros Supplier No. 117241000
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2524-64-3 | C12H10ClO3P | 268.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.63
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 314°C to 316°C (272.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5O)2P(O)Cl
Refractive Index 1.5485 to 1.5505
Quantity 100 mL
Beilstein 03, 179
Fieser 01,345; 02,180; 03,133; 11,223
Merck Index 15, 3367
Specific Gravity 1.29
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Viscosity 14 mPa.s (25°C)
Formula Weight 268.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenyl chlorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyl chlorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.