missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl cyanocarbonimidate, 97%

Product Code. 11407947
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407947 1 g 1g
11417947 5 g 5g
11427947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407947 Supplier Thermo Scientific Alfa Aesar Supplier No. A16600.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 79463-77-7 | C14H10N2O2 | 238.25 g/mol

Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep under a nitrogen blanket. Store away from strong oxidizing agents, strong acids, strong bases, moist air, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79463-77-7
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.25
MDL Number MFCD00010380
InChI Key SLIKWVTWIGHFJE-UHFFFAOYSA-N
Synonym diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile
PubChem CID 688090
IUPAC Name cyano(diphenoxymethylidene)amine
SMILES N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 155°C to 158°C
Linear Formula (C6H5O)2C=NCN
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 238.25
Percent Purity 97%
Chemical Name or Material Diphenyl cyanocarbonimidate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.